Please login to your online account to display your discounted pricing
Mott Manufacturing Steel Casework Specialty Cabinet, Acid Storage Units, Single Door
Chemically resistant molded liner and one black phenolic shelf for acid storage
$1,055.35 - $1,081.95
Products
Catalog Number Mfr. No. Price Quantity    
36-104-1442
mott lab inc
1111011
Each for $1,055.35
36-104-1443
mott lab inc
1111211
Each for $1,055.35
36-104-1444
mott lab inc
1211011
Each for $1,081.95
36-104-1445
mott lab inc
1211211
Each for $1,081.95
36-104-1446
mott lab inc
1231011
Each for $1,062.44
36-104-1447
mott lab inc
1231211
Each for $1,062.44
Description & Specifications
  • Furniture grade cold rolled steel with high pre and post consumer recycled content
  • Laboratory grade powder coating - Warm Gray 601006
  • Contemporary inset design
  • Brushed aluminum bar handle(s) 10.1cm (4 in.)
  • Five-knuckle stainless steel hinges and four levelling feet
  • One piece molded polyethylene liner with 2.54cm (1 in.) raised front edge to contain spills
  • Single black phenolic adjustable shelf
  • Full height removable back panel(s)
  • Door(s) with double wall construction enclosed with sound deadening material
  • Interior door lined with chemically resistant FRP and plastic roller catch(es)
  • Threaded vent connection, fusion welded to cabinet with standard plastic pipe fittings
Meets SEFA 8 performance standards