Please login to your online account to display your discounted pricing
Mott Manufacturing Steel Casework Component, Apron Units
Two piece adjustable construction
$191.56 - $205.75
Products
Catalog Number Mfr. No. Price Quantity    
36-104-1401
mott lab inc
AUC0130
Each for $191.56
36-104-1402
mott lab inc
AUC0136
Each for $205.75
Description & Specifications
bullets1 (begin)Furniture grade cold rolled steel with high pre and post consumer recycled content
Aprons provide a work area from a seated position
Available with or without drawers, 38.1cm (15 in.) wide drawer
Use option code “OL” when ordering Architectural Overlay design
Laboratory grade powder coating - Warm Gray 601006bullets1 (end)
Meets SEFA 8 performance standards