Please login to your online account to display your discounted pricing
Mott Manufacturing Steel Casework Component, Apron Service Covers
Two piece construction with mounting angles
$140.12 - $148.98
Products
Catalog Number Mfr. No. Price Quantity    
36-104-1405
mott lab inc
SCP3030
Each for $140.12
36-104-1406
mott lab inc
SCP3036
Each for $148.98
Description & Specifications
  • Furniture grade cold rolled steel with high pre and post consumer recycled content
  • Apron service cover panels are used to conceal open spaces and services below aprons
  • Attaches to adjacent cabinets and meets bottom of apron
  • Assembly includes single angles for attachment to adjacent cabinets
  • Removable Upper panel to allow access to services
  • Fixed lower panel in order to apply coved base and overlapping lower and upper panel allows to accommodate for uneven floors
  • Available for sitting height 63.5cm (25 in.), ADA height 71.1cm (28 in.) and standing height 78.7cm (31 in.) applications
  • Laboratory grade powder coating - Warm Gray 601006
Meets SEFA 8 performance standards