Please login to your online account to display your discounted pricing
Mott Manufacturing Altus™ Table System Component, Assembly
Workstation with welded frame and with adjustable height
$1,284.14 - $1,340.91
Products
Catalog Number Mfr. No. Price Quantity    
36-104-1408
mott lab inc
ATS2348
Each for $1,284.14
36-104-1409
mott lab inc
ATS2360
Each for $1,314.30
36-104-1410
mott lab inc
ATS2372
Each for $1,340.91
Description & Specifications
  • Furniture grade cold rolled steel with high pre and post consumer recycled content
  • Laboratory grade powder coating - Warm Gray 601006
  • Overall assembly is 213.3cm (84 in.) high, 5.08 x 7.62cm (2 x 3 in.) deep rear posts, 5.08cm (2 in.) square tubular front legs with leveling feet
  • All welded construction and height adjustable cantilevered table frame in 2.54cm (1 in.) increments from 73.6 to 91.4cm (29 to 36 in.) (work surface not included)
  • Complete with handing rails to suspend lower cabinets (cabinets not included)
  • Uprights punched to accept shelf brackets, adjustable in 2.54cm (1 in.) increments (shelf brackets not included)
  • Punched for maximum of three fixtures on plumbing post (fixtures and plumbing not included)
  • Three prewired duplex electrical receptacles on electrical post
  • Will support 272.1kg (600 lb.) load, evenly distributed. Ships partially assembled
  • Countertop not included (overall depth of assembly does not include worksurface)
  • Approved to UL 962 standards
Meets SEFA 8 performance standards