Please login to your online account to display your discounted pricing

Alfa Aesar™ Motilin, Human, Porcine

For cellular and molecular biology applications

$174.78 - $174.78

Specifications

Product Type Motilin, human, porcine
Cross Reactivity Human, Porcine
Molecular Weight 2714.10
Storage Requirements -30° to -10°C
Sequence Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Met-Gln-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66400MCR

 
alfa aesar™
J66400MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66400EXD

 
alfa aesar™
J66400EXD
0.5mg Each for $174.78
Description & Specifications

Specifications

Product Type Motilin, human, porcine
Cross Reactivity Human, Porcine
Molecular Weight 2714.10
Storage Requirements -30° to -10°C
Sequence Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Met-Gln-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln
Format Solid
Regulatory Status RUO
Purification Method HPLC