Please login to your online account to display your discounted pricing
National Optical Moticam™ Digital Microscope Camera
Use your own microscope to capture high-resolution still or video images with the Moticam Digital Microscope Camera.
Manufacturer: national optical  MOTICAM2
 View more versions of this product
Catalog No. S02359A
  • / Each
Description & Specifications
Specifications
Resolution 2 MP digital signal @ 1600x1200 maximum resolution
Includes CS Ring Adaptor, Focusable Lens, 30mm and 38mm Eyepiece Adapters, USB3.0 Cable, Calibration Slide, Macro Tube, Motic Images Plus 3.0 for PC and OSX
Compatibility (Operating System) Windows XP/Vista/7/Apple Mac OS
Required Accessories Processor: 2 Ghz or Above, USB 2.0 (minimum requirement), USB 3.0 (highly recommended), RAM: 2 GB, Hard disk: 1 GB (free space), Video card: 128 MB video memory
For Use With (Equipment) Microscopes
Item Description Digital 2MP Microscope Camera
Warranty 1 Year
Sensor Types CMOS
Product Type Digital microscope camera
  • High-resolution camera permits you to use your own microscope to create still or moving microscope images on your PC
  • Lightweight digital camera mounts over almost any microscope eyepiece (stereo or compound) with supplied C ring adapter and microscope eyepiece adapters
  • Included Motic Images Plus software allows user to view, enhance, label, measure, print, and store the images all with one program