Fisher Science Education™ Moticam™ Digital Microscope Camera
Use your own microscope to capture high-resolution still or video images with the Moticam Digital Microscope Camera.
Manufacturer:  Fisher Science Education™ MOTICAM5PLUS
 View more versions of this product
Catalog No. S22241
Description
Description
  • High-resolution camera permits you to use your own microscope to create still or moving microscope images on your PC
  • Lightweight digital camera mounts over almost any microscope eyepiece (stereo or compound) with supplied C ring adapter and microscope eyepiece adapters
  • Included Motic Images Plus software allows user to view, enhance, label, measure, print, and store the images all with one program
Specifications
Specifications
Digital 5MP USB 3.0 Microscope Camera
Digital microscope camera
Windows XP/Vista/7/Apple Mac OS
Processor: 2 Ghz or Above, USB 2.0 (minimum requirement), USB 3.0 (highly recommended), RAM: 2 GB, Hard disk: 1 GB (free space), Video card: MUST have dedicated 256MB video memory
CMOS
1 Year
Microscopes
CS Ring Adaptor, Focusable Lens, 30mm and 38mm Eyepiece Adapters, USB3.0 Cable, Calibration Slide, Macro Tube, Motic Images Plus 3.0 for PC and OSX
5 MP digital signal @ 2592x1944 maximum resolution
Documents