missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Morpholinoacetonitrile, TCI America™

Manufacturer:  TCI America M10955G

 View more versions of this product

Catalog No. M10955G



Specifications

Specifications

Morpholinoacetonitrile
White
5g
C6H10N2O
3439
OOSOCAXREAGIGA-UHFFFAOYSA-N
2-morpholin-4-ylacetonitrile
22055
Crystal-Powder at 20°C
124°C
62°C
5807-02-3
MFCD00023372
N-Cyanomethylmorpholine
C1COCCN1CC#N
126.16
126.16
≥98.0% (GC,T)

Chemical Identifiers

5807-02-3
126.16
OOSOCAXREAGIGA-UHFFFAOYSA-N
22055
C1COCCN1CC#N
C6H10N2O
MFCD00023372
N-Cyanomethylmorpholine
2-morpholin-4-ylacetonitrile
Safety and Handling

Safety and Handling

RTECSNumber : QD6650000

TSCA : No

Documents