Please login to your online account to display your discounted pricing

Morpholine Standard, Certified, 1000ppm ±10ppm (1mL = 1mg), LabChem™

$129.20 - $129.20

Chemical Identifiers

Chemical Name or Material Morpholine Standard
CAS 7732-18-5
CAS Max % 99.9
CAS 110-91-8
CAS Max % 0.1
Name Note 1000ppm (1mL = 1mg)
Purity Grade Certified Grade
Linear Formula C4H9NO
Formula Weight 87.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC174501

 
lab chem
LC174501
Poly Bottle 500mL Each for $129.20
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 87.12
Chemical Name or Material Morpholine Standard
Color Clear, colorless
Quantity 500mL
Name Note 1000ppm (1mL = 1mg)
Physical Form Liquid
Concentration 1000ppm ±10ppm (1mL = 1mg)
Linear Formula C4H9NO
CAS 7732-18-5
Identification Passes Test
CAS Max % 99.9
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 110-91-8
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
Specific Gravity 1g/mL
CAS Max % 0.1
Recommended Storage Room Temperature
Density 1g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature