Please login to your online account to display your discounted pricing

Morpholine, ACS Grade, ≥ 99.0 %, LabChem™

$100.37 - $100.37

Chemical Identifiers

Chemical Name or Material Morpholine
CAS 110-91-8
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C4H9NO
Formula Weight 87.12
UN Number UN2054
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC174401

 
lab chem
LC174401
Amber Glass 500mL Each for $100.37
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 87.12
Chemical Name or Material Morpholine
EINECS Number 203-815-1
Density 0.99g/cm3
Identification Passes Test
Purity Grade ACS Grade
Physical Form Liquid
Linear Formula C4H9NO
CAS 110-91-8
UN Number UN2054
Packaging Amber Glass
Health Hazard 1 Danger
Solubility Soluble in water
Specific Gravity 0.99g/cm3
CAS Max % 100
Health Hazard 2 GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use alcohol resistant foam, carbon dioxide, sand to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; May release flammable gases; Thermal decomposition generates: Corrosive vapors
Boiling Point 128°C
Melting Point −5°C
Color Clear, colorless
Flash Point 32°C
Quantity 500mL
Recommended Storage Room Temperature
Safety and Handling

Danger

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use alcohol resistant foam, carbon dioxide, sand to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life.

Room Temperature