Learn More
Morpholine, ACS, 99.0% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03198436
Description
Morpholine is used to make other chemicals, as a corrosion inhibitor, and in detergents. It is also used as solvent and optical brightener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Morpholine | |
-5°C | |
1.007 g/mL | |
35°C (95°F) | |
C4H9NO | |
MFCD00005972 | |
UN2054 | |
Hygroscopic | |
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro | |
YNAVUWVOSKDBBP-UHFFFAOYSA-N | |
morpholine | |
8083 | |
87.12 | |
ACS |
110-91-8 | |
Colorless | |
129°C | |
Amine-like | |
1.454 | |
500 g | |
102549 | |
14,6277 | |
Soluble in water. | |
C1COCCN1 | |
87.12 | |
CHEBI:34856 | |
≤99.0% | |
Liquid |
Chemical Identifiers
110-91-8 | |
87.12 | |
YNAVUWVOSKDBBP-UHFFFAOYSA-N | |
8083 | |
morpholine |
C4H9NO | |
MFCD00005972 | |
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro | |
CHEBI:34856 | |
C1COCCN1 |
Safety and Handling
GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302-H311+H331-H314-H335-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: MORPHOLINE
EINECSNumber : 203-815-1
RTECSNumber : QD6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only