missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Morpholine, 99+%, extra pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC158680100

 View more versions of this product

Catalog No. AC158680100



Specifications

Specifications

Morpholine
32°C
Plastic drum
0.99
126.0°C to 130.0°C
-5.0°C
110-91-8
100.0
C4H9NO
27, 5
15, 6362
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil
C1COCCN1
87.12
CHEBI:34856
Liquid
Extra Pure
0.9900g/mL
Authentic
1.4530 to 1.4550
2.23 mPa.s (20°C)
Undesignated
10L
99.0
99% min. (GC)
MFCD00005972
01,705; 11,352; 13,14
1-oxa-4-azacyclohexane
YNAVUWVOSKDBBP-UHFFFAOYSA-N
morpholine
8083
87.12
99+%

Chemical Identifiers

110-91-8
87.12
YNAVUWVOSKDBBP-UHFFFAOYSA-N
8083
morpholine
C4H9NO
MFCD00005972
1-oxa-4-azacyclohexane
CHEBI:34856
C1COCCN1
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (

GHS Signal Word: Danger

EINECSNumber : 203-815-1

SDS Documents