Learn More
Morpholine, 99.5%, purified by redistillation, AcroSeal™, Thermo Scientific Chemicals
$138.24 - $138.24
Chemical Identifiers
CAS | 110-91-8 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.12 |
MDL Number | MFCD00005972 |
InChI Key | YNAVUWVOSKDBBP-UHFFFAOYSA-N |
Synonym | 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro |
PubChem CID | 8083 |
ChEBI | CHEBI:34856 |
IUPAC Name | morpholine |
SMILES | C1COCCN1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC433851000
|
Thermo Scientific Chemicals
433851000 |
100 mL | AcroSeal™ Glass bottle |
Each for $138.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-91-8 | |
87.12 | |
YNAVUWVOSKDBBP-UHFFFAOYSA-N | |
8083 | |
morpholine |
C4H9NO | |
MFCD00005972 | |
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro | |
CHEBI:34856 | |
C1COCCN1 |
Specifications
110-91-8 | |
0.9900g/mL | |
32°C | |
AcroSeal™ Glass bottle | |
1.4530 to 1.4550 | |
100 mL | |
15,6362 | |
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil | |
C1COCCN1 | |
87.12 | |
2.23 mPa.s (20°C) | |
87.12 | |
Morpholine |
-5.0°C | |
126.0°C to 130.0°C | |
99.4% min. (GC) | |
C4H9NO | |
MFCD00005972 | |
0.99 | |
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro | |
YNAVUWVOSKDBBP-UHFFFAOYSA-N | |
morpholine | |
8083 | |
CHEBI:34856 | |
99.5% |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
GHS Signal Word: Danger
EINECSNumber : 203-815-1
RUO – Research Use Only