missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (±)-Morpholine-3-carboxylic acid hydrochloride, 96%

$78.19 - $286.84

Chemical Identifiers

CAS 77873-76-8
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD01318649
InChI Key JUNOWSHJELIDQP-UHFFFAOYSA-N
Synonym 3-morpholinecarboxylic acid, 3-morpholine carboxylic acid, r-3-morpholinecarboxylic acid, 3-morpholinecarboxylicacid, 3-morpholinecarboxylicacid, 3r, 3-morpholinecarboxylicacid, 3s, acmc-20agwb, acmc-209pcc, acmc-209vok, 3-morphline carboxylic acid
PubChem CID 3340152
IUPAC Name morpholine-3-carboxylic acid
SMILES C1COCC(N1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64336MD View Documents Alfa Aesar™
H64336MD
250mg Each for $78.19
 
AAH6433603 View Documents Alfa Aesar™
H6433603
1g Each for $286.84
 
Specifications

Chemical Identifiers

77873-76-8
131.131
JUNOWSHJELIDQP-UHFFFAOYSA-N
3340152
C1COCC(N1)C(=O)O
C5H9NO3
MFCD01318649
3-morpholinecarboxylic acid, 3-morpholine carboxylic acid, r-3-morpholinecarboxylic acid, 3-morpholinecarboxylicacid, 3-morpholinecarboxylicacid, 3r, 3-morpholinecarboxylicacid, 3s, acmc-20agwb, acmc-209pcc, acmc-209vok, 3-morphline carboxylic acid
morpholine-3-carboxylic acid

Specifications

White
250mg
C5H9NO3
UN2811
3-morpholinecarboxylic acid, 3-morpholine carboxylic acid, r-3-morpholinecarboxylic acid, 3-morpholinecarboxylicacid, 3-morpholinecarboxylicacid, 3r, 3-morpholinecarboxylicacid, 3s, acmc-20agwb, acmc-209pcc, acmc-209vok, 3-morphline carboxylic acid
C1COCC(N1)C(=O)O
131.131
167.59
96%
245°C to 250°C
77873-76-8
MFCD01318649
115879
JUNOWSHJELIDQP-UHFFFAOYSA-N
morpholine-3-carboxylic acid
3340152
Powder
(±)-Morpholine-3-carboxylic acid hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.