Please login to your online account to display your discounted pricing

Morpholin-2-ylmethyl-carbamic acid tert-butyl ester, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO08372DA

 View more versions of this product

Catalog No. MO08372DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 216.28
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Chemical Name or Material Morpholin-2-ylmethyl-carbamic acid tert-butyl ester
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
MDL Number MFCD06808588
Packaging Amber Glass Bottle
Molecular Formula C10H20N2O3
CAS 173341-02-1

Specifications

Chemical Name or Material Morpholin-2-ylmethyl-carbamic acid tert-butyl ester
CAS 173341-02-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H20N2O3
Formula Weight 216.28
MDL Number MFCD06808588
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents