missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Morpholin-2-ylmethyl-carbamic acid tert-butyl ester, 97%, Maybridge

$331.11 - $331.11

Chemical Identifiers

CAS 173341-02-1
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C10H20N2O3
Formula Weight 216.28
MDL Number MFCD06808588
Chemical Name or Material Morpholin-2-ylmethyl-carbamic acid tert-butyl ester
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MO08372DA SDS Fisher Scientific Company
MO08372DA
Amber Glass Bottle 1g Each for $331.11
Specifications Description & Specifications

Chemical Identifiers

173341-02-1
97%
216.28
Morpholin-2-ylmethyl-carbamic acid tert-butyl ester
97.0
C10H20N2O3
MFCD06808588
97%

Specifications

173341-02-1
97%
216.28
Amber Glass Bottle
Morpholin-2-ylmethyl-carbamic acid tert-butyl ester
97.0
C10H20N2O3
MFCD06808588
1g
97%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger