Please login to your online account to display your discounted pricing

Morphix Technologies Color Comparators for ChromAir™ Passive Sampling Systems

Use with ChromAir badge to determine the exact dose of a specific contaminant present in the workplace.

$124.87 - $132.97

Specifications

For Use With (Application) To determine the exact dose of a specific contaminant present in the workplace
Measurement Range 0.3 to 40ppm.Hour
View More Specs
Products
Catalog Number Mfr. No. Detectable Analytes Price Quantity    

18-999-1389

 
morphix technologies
384003
Mercury Each for $132.97

18-999-1386

 
morphix technologies
384001
Formaldehyde Each for $124.87
Description & Specifications

Specifications

For Use With (Application) To determine the exact dose of a specific contaminant present in the workplace
Measurement Range 0.3 to 40ppm.Hour

Color scale identically matches the colors formed on a ChromAir badge when it is exposed to a contaminant.

  • Badge slides in comparator for comparison to color scale insert
  • Exact exposure dose is shown in the exposure dose window