Morphix Technologies ChromAir™ Passive Sampling Badges DFS Item
Built-in scale features six dose ranges. Morphix Technologies ChromAir Passive Sampling Quantitative Badges provide a cost-effective means of sampling for gases and vapors and are ideal for personnel and area monitoring.
Manufacturer:  Morphix Technologies 38002010
 View more versions of this product
Catalog No. 19-087530
Description
Description
  • Built-in scale for direct reading of exposure dose features six dose ranges that change color upon exposure
  • Color change and exposure measurement are easy to read, providing fast, accurate, onsite results
  • Badge's patented design minimizes the effects of temperature, humidity, face velocity, and interferences
Order Info
Attaches to user's collar with badge clip (19-087-526, sold separately–contact your Fisher Safety Customer Service Representative for ordering information). Available 10 badges per pack.
Specifications
Specifications
Acetone
20 to 24000ppm.Hour
1 Day
10 to 45°C
0.01kg
The ChromAir™ Acetone badge may be used for personnel or area monitoring for exposure times ranging from 1 min. to 24 Hours
Sampling badge
50 to 113°F
0.02 lb.