Please login to your online account to display your discounted pricing

Morin Solution, LabChem™

$75.37 - $75.37

Chemical Identifiers

Chemical Name or Material Morin Solution
CAS 67-56-1
CAS Max % 95
CAS 654055-01-3
CAS Max % 5
Linear Formula C15H10O7
Formula Weight 302.24
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC174207

 
lab chem
LC174207
125mL Each for $75.37
Description & Specifications

Specifications

Formula Weight 302.24
Chemical Name or Material Morin Solution
Decomposition Information Carbon monoxide; Carbon dioxide
Color Dark brown
Quantity 125mL
Physical Form Liquid
Linear Formula C15H10O7
CAS 67-56-1
UN Number UN1993
Health Hazard 1 Danger
CAS Max % 95
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, liver, kidneys, optic nerve).
Identification Passes Test
CAS 654055-01-3
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 5
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, liver, kidneys, optic nerve).