missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Morin Hydrate, TCI America™

Manufacturer:  TCI America P004125G

 View more versions of this product

Catalog No. P004125G



Specifications

Specifications

Morin Hydrate
25g
C15H12O8
3077
MYUBTSPIIFYCIU-UHFFFAOYSA-N
2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
16219651
Crystal-Powder at 20°C
Undesignated
654055-01-3
MFCD00006826
morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin hydrate, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
C1=CC(=C(C=C1O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O.O
320.253
302.24
≥90.0% (HPLC)

Chemical Identifiers

654055-01-3
320.253
MYUBTSPIIFYCIU-UHFFFAOYSA-N
16219651
C1=CC(=C(C=C1O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O.O
C15H12O8
MFCD00006826
morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin hydrate, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5230

RTECSNumber : LK8749000

TSCA : Yes

Documents