Please login to your online account to display your discounted pricing

Morin hydrate, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC354000500

 View more versions of this product

Catalog No. AC354000500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 302.24
Physical Form Powder
Quantity 50g
Color Yellow to brown
TSCA TSCA
EINECS Number 207-542-9
RTECS Number LK8749000
Beilstein 18, 239
Merck Index 14, 6269
Water 7% max.
UV Spectrometry Complies
Packaging Glass bottle
Health Hazard 2 GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. Harmful if swallowed. Suspected of causing cancer.
Health Hazard 1 Warning
CAS 480-16-0
Sulfated Ash 2% max.
Infrared Spectrum Authentic
Synonym 2',3,4',5,7-Pentahydroxyflavone hydrate
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C15H10O7·xH2O
Chemical Name or Material Morin hydrate
MDL Number MFCD00217054
CAS 654055-01-3

Specifications

Chemical Name or Material Morin hydrate
CAS 654055-01-3
CAS 480-16-0
Molecular Formula C15H10O7·xH2O
Formula Weight 302.24
MDL Number MFCD00217054
Synonym 2',3,4',5,7-Pentahydroxyflavone hydrate
Accessories

Warning

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. Harmful if swallowed. Suspected of causing cancer.

SDS & Safety Documents