Please login to your online account to display your discounted pricing

Morin hydrate, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC354000050

 View more versions of this product

Catalog No. AC354000050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 302.24
Quantity 5g
Color Yellow to brown
Beilstein 18, 239
Merck Index 14, 6269
EINECS Number 207-542-9
TSCA TSCA
RTECS Number LK8749000
CAS 480-16-0
Water 7% max.
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Sulfated Ash 2% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 2',3,4',5,7-Pentahydroxyflavone hydrate
UV Spectrometry Complies
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. Harmful if swallowed. Suspected of causing cancer.
Molecular Formula C15H10O7·xH2O
CAS 654055-01-3
Chemical Name or Material Morin hydrate
MDL Number MFCD00217054

Specifications

Chemical Name or Material Morin hydrate
CAS 654055-01-3
CAS 480-16-0
Molecular Formula C15H10O7·xH2O
Formula Weight 302.24
MDL Number MFCD00217054
Synonym 2',3,4',5,7-Pentahydroxyflavone hydrate
Accessories

Warning

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. Harmful if swallowed. Suspected of causing cancer.

SDS & Safety Documents