Please login to your online account to display your discounted pricing

Mordant Blue 13 dihydrate, 99%, pure, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  415140050

 View more versions of this product

Catalog No. AC415140050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 554.85
Quantity 5g
EINECS Number 213-894-4
TSCA TSCA
Purity Grade Pure
Synonym 3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxy-2,7-naphthalene disulfonic acid, disodium salt
Chemical Name or Material Mordant Blue 13 dihydrate
Molecular Formula C16 H9 Cl N2 Na2 O9 S2 . 2 H2 O
CAS 1058-92-0
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
MDL Number MFCD00003941

Specifications

Chemical Name or Material Mordant Blue 13 dihydrate
Name Note 99%
CAS 1058-92-0
Purity Grade Pure
Assay Percent Range 99%
Molecular Formula C16 H9 Cl N2 Na2 O9 S2 . 2 H2 O
Formula Weight 554.85
MDL Number MFCD00003941
Synonym 3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxy-2,7-naphthalene disulfonic acid, disodium salt