Please login to your online account to display your discounted pricing

Mordant Blue 13 dihydrate, 99%, pure, ACROS Organics™

$57.71 - $57.71

Chemical Identifiers

Chemical Name or Material Mordant Blue 13 dihydrate
Name Note 99%
CAS 1058-92-0
Purity Grade Pure
Assay Percent Range 99%
Molecular Formula C16 H9 Cl N2 Na2 O9 S2 . 2 H2 O
Formula Weight 554.85
MDL Number MFCD00003941
Synonym 3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxy-2,7-naphthalene disulfonic acid, disodium salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC415140050

  SDS
thermo fisher scientific
415140050
Glass bottle 5g Each for $57.71
Description & Specifications

Specifications

Formula Weight 554.85
Quantity 5g
TSCA TSCA
EINECS Number 213-894-4
Purity Grade Pure
Synonym 3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxy-2,7-naphthalene disulfonic acid, disodium salt
Packaging Glass bottle
Chemical Name or Material Mordant Blue 13 dihydrate
CAS 1058-92-0
Molecular Formula C16 H9 Cl N2 Na2 O9 S2 . 2 H2 O
MDL Number MFCD00003941
Assay Percent Range 99%
Name Note 99%