missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPSO, 98%

Biological buffers, pH range 6.2 to 7.6

Manufacturer:  Alfa Aesar™ 4413518

 View more versions of this product

Catalog No. AA4413518



Specifications

Specifications

MOPSO
50g
C7H15NO5S
2-hydroxy-3-morpholin-4-yl propane-1-sulfonic acid
NUFBIAUZAMHTSP-UHFFFAOYSA-N
2-hydroxy-3-morpholin-4-ylpropane-1-sulfonic acid
109333
Powder
276°C to 279°C
68399-77-9
MFCD00009629
Water solubility under desired conditions ca.112,6g/L at 20°C.
C1COCCN1CC(CS(=O)(=O)O)O
225.259
225.27
98%

Chemical Identifiers

68399-77-9
225.259
NUFBIAUZAMHTSP-UHFFFAOYSA-N
109333
C1COCCN1CC(CS(=O)(=O)O)O
C7H15NO5S
MFCD00009629
2-hydroxy-3-morpholin-4-yl propane-1-sulfonic acid
2-hydroxy-3-morpholin-4-ylpropane-1-sulfonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 269-989-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents