missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPSO, 0.2M buffer soln., pH 6.5

$84.96 - $133.08

Chemical Identifiers

CAS 79803-73-9
Molecular Formula C7H14NNaO5S
Molecular Weight (g/mol) 247.241
MDL Number MFCD00070007
InChI Key WSFQLUVWDKCYSW-UHFFFAOYSA-M
PubChem CID 23675131
IUPAC Name sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61833AE View Documents Alfa Aesar™
J61833AE
100mL Each for $84.96
 
AAJ61833AK View Documents Alfa Aesar™
J61833AK
250mL Each for $133.08
 
Specifications

Chemical Identifiers

79803-73-9
247.241
WSFQLUVWDKCYSW-UHFFFAOYSA-M
sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate
C7H14NNaO5S
MFCD00070007
23675131
C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+]

Specifications

79803-73-9
MFCD00070007
WSFQLUVWDKCYSW-UHFFFAOYSA-M
sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate
23675131
6.5
MOPSO buffer soln.
C7H14NNaO5S
0.2M buffer soln.
C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+]
247.241
Liquid
100mL