Please login to your online account to display your discounted pricing

MOPS sodium salt, 98%, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC352590010

 View more versions of this product

Catalog No. AC352590010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 231.25
Assay ≥97.5%
Quantity 1kg
Color White
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 97.5
Packaging Glass bottle
Synonym 3-(4-Morpholino)propanesulfonic acid sodium salt
Chemical Name or Material MOPS sodium salt
MDL Number MFCD00064350
CAS 71119-22-7
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H14NNaO4S

Specifications

Chemical Name or Material MOPS sodium salt
Name Note 98%
CAS 71119-22-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H14NNaO4S
Formula Weight 231.25
MDL Number MFCD00064350
Synonym 3-(4-Morpholino)propanesulfonic acid sodium salt