Please login to your online account to display your discounted pricing

MOPS, Free Acid, Molecular Biology Grade, Calbiochem™, EMD Millipore

A zwitterionic buffer useful in the pH range of 6.5 to 7.9

$86.00 - $86.00

Chemical Identifiers

Assay Percent Range ≥99.0%
Formula Weight 209.3
Buffers MOPS, Free Acid, Molecular Biology Grade
CAS 1132-61-2
Chemical Name or Material MOPS, Free Acid
Molecular Formula C7H15NO4S
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

475922100GM

 
emd millipore corporation
475922100GM
Each for $86.00
Description & Specifications

Specifications

Formula Weight 209.3
Quantity 100g
Chemical Name or Material MOPS, Free Acid
Buffers MOPS, Free Acid, Molecular Biology Grade
Purity Grade Notes Potentiometry
Assay Percent Range ≥99.0%
Molecular Formula C7H15NO4S
CAS 1132-61-2
DNase None Detected
Synonym 3-(N-Morpholino)propanesulfonic Acid
Solubility Information Soluble in cold water
Protease None Detected
Purity Molecular Biology Grade
Recommended Storage 15°C to 30°C (Do not freeze).
RNase None Detected
Safety and Handling

15°C to 30°C (Do not freeze).