Please login to your online account to display your discounted pricing

MOPS/EDTA Buffer, 10X Liquid Concentrate, Molecular Biology Grade, Calbiochem™, EMD Millipore

Prepared with DEPC for RNA applications

$113.00 - $113.00

Chemical Identifiers

Analyte or Component Names MOPS, Sodium Acetate, EDTA
Buffers MOPS/EDTA Buffer
Chemical Name or Material MOPS/EDTA Buffer
Concentration or Composition (by Analyte or Components) 10X concentrate contains 200mM MOPS, 50mM sodium acetate, 10mM EDTA
Molecular Formula C7H15NO4S
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

475916100ML

 
emd millipore corporation
475916100ML
Each for $113.00
Description & Specifications

Specifications

Quantity 100mL
Chemical Name or Material MOPS/EDTA Buffer
Analyte or Component Names MOPS, Sodium Acetate, EDTA
Buffers MOPS/EDTA Buffer
Molecular Formula C7H15NO4S
Concentration or Composition (by Analyte or Components) 10X concentrate contains 200mM MOPS, 50mM sodium acetate, 10mM EDTA
Protease None Detected
Purity Molecular Biology Grade
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
RNase None Detected
DNase None Detected
Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).