Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPS-buffered saline (5X, pH 7.4), low endotoxin


Quantity 100mL

Manufacturer: alfa aesar™  J67506AE

 View more versions of this product

Catalog No. AAJ67506AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material MOPS-buffered saline (5X, pH 7.4)
Physical Form Liquid
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin

Specifications

Chemical Name or Material MOPS-buffered saline (5X, pH 7.4)
Name Note low endotoxin