missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPS, 1.0M buffer soln., pH 6.0

$130.46 - $200.47

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62840AK View Documents Alfa Aesar™
J62840AK
250mL Each for $130.46
 
AAJ62840AP View Documents Alfa Aesar™
J62840AP
500mL Each for $200.47
 
Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
CHEBI:44115
C1COCCN1CCCS(=O)(=O)O
C7H15NO4S
MFCD00006183
70807
3-morpholin-4-ylpropane-1-sulfonic acid

Specifications

1132-61-2
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
Liquid
6.0
250mL
C7H15NO4S
1.0M buffer soln.
C1COCCN1CCCS(=O)(=O)O
209.26
CHEBI:44115
Odorless
Colorless
MOPS buffer soln.
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA