missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPS, 0.5M buffer soln., pH 8.5

$74.16 - $111.18

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62368AE View Documents Alfa Aesar™
J62368AE
100mL Each for $74.16
 
AAJ62368AK View Documents Alfa Aesar™
J62368AK
250mL Each for $111.18
 
Description

Description

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
CHEBI:44115
C1COCCN1CCCS(=O)(=O)O
C7H15NO4S
MFCD00006183
70807
3-morpholin-4-ylpropane-1-sulfonic acid

Specifications

1132-61-2
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
Liquid
8.5
MOPS buffer soln.
C7H15NO4S
0.5M buffer soln.
C1COCCN1CCCS(=O)(=O)O
209.26
CHEBI:44115
Odorless
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C