missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPS, 0.5M buffer soln., pH 8.0

$78.73 - $117.69

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61236AE View Documents Alfa Aesar™
J61236AE
100mL Each for $78.73
 
AAJ61236AK View Documents Alfa Aesar™
J61236AK
250mL Each for $117.69
 
Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
CHEBI:44115
C1COCCN1CCCS(=O)(=O)O
C7H15NO4S
MFCD00006183
70807
3-morpholin-4-ylpropane-1-sulfonic acid

Specifications

1132-61-2
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
Liquid
8.0
MOPS buffer soln.
C7H15NO4S
0.5M buffer soln.
C1COCCN1CCCS(=O)(=O)O
209.26
CHEBI:44115
Odorless
100mL
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures