missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPS, 0.5M buffer soln., pH 8.0

Manufacturer:  Alfa Aesar™ J61236AK

 View more versions of this product

Catalog No. AAJ61236AK



Specifications

Specifications

1132-61-2
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
Liquid
8.0
C7H15NO4S
0.5M buffer soln.
C1COCCN1CCCS(=O)(=O)O
209.26
CHEBI:44115
Odorless
250mL

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
CHEBI:44115
C1COCCN1CCCS(=O)(=O)O
C7H15NO4S
MFCD00006183
70807
3-morpholin-4-ylpropane-1-sulfonic acid
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents