missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MOPS, 0.2M buffer soln., pH 7.4, low endotoxin, Thermo Scientific™
$101.81 - $101.81
Chemical Identifiers
CAS | 1132-61-2 |
---|---|
Molecular Formula | C7H15NO4S |
Molecular Weight (g/mol) | 209.26 |
MDL Number | MFCD00006183 |
InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
PubChem CID | 70807 |
ChEBI | CHEBI:44115 |
IUPAC Name | 4-(3-sulfonatopropyl)morpholin-4-ium |
SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67497AE
|
Thermo Scientific Chemicals
J67497AE |
100 mL |
Each for $101.81
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1132-61-2 | |
209.26 | |
DVLFYONBTKHTER-UHFFFAOYSA-N | |
CHEBI:44115 | |
[O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
C7H15NO4S | |
MFCD00006183 | |
70807 | |
4-(3-sulfonatopropyl)morpholin-4-ium |
Specifications
1132-61-2 | |
0.2 M | |
Liquid | |
C7H15NO4S | |
DVLFYONBTKHTER-UHFFFAOYSA-N | |
4-(3-sulfonatopropyl)morpholin-4-ium | |
70807 | |
MOPS, low endotoxin |
Colorless | |
7.4 | |
100 mL | |
MFCD00006183 | |
[O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
209.26 | |
CHEBI:44115 |
Safety and Handling
H335
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold