Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPS, 0.2M buffer soln., pH 6.5, low endotoxin


Quantity 100mL

Manufacturer: alfa aesar™  J67474AE

 View more versions of this product

Catalog No. AAJ67474AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material MOPS
MDL Number MFCD00006183
Physical Form Liquid
TSCA TSCA
Name Note low endotoxin
Storage Note 1 2° to 8°C
pH 6.5
Color Colorless
Concentration 0.2M buffer soln.
Quantity 100mL
CAS 1132-61-2

Specifications

Chemical Name or Material MOPS
CAS 1132-61-2
Name Note low endotoxin
MDL Number MFCD00006183
Accessories

2° to 8°C