Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPS, 0.2M buffer soln., pH 6.5, low endotoxin

Chemical Identifiers

Chemical Name or Material MOPS
CAS 1132-61-2
Name Note low endotoxin
MDL Number MFCD00006183
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67474AE

 
alfa aesar™
J67474AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67474AK

 
alfa aesar™
J67474AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material MOPS
MDL Number MFCD00006183
Physical Form Liquid
TSCA TSCA
Name Note low endotoxin
Storage Note 1 2° to 8°C
pH 6.5
Color Colorless
Concentration 0.2M buffer soln.
Quantity 100mL
CAS 1132-61-2
Safety and Handling

2° to 8°C