Please login to your online account to display your discounted pricing

Alfa Aesar™ MOPS, 0.2M buffer soln., pH 6.5, low endotoxin


Quantity 250mL

Manufacturer: alfa aesar™  J67474AK

 View more versions of this product

Catalog No. AAJ67474AK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material MOPS
Physical Form Liquid
CAS 1132-61-2
pH 6.5
Color Colorless
Quantity 250mL
TSCA TSCA
Name Note low endotoxin
Concentration 0.2M buffer soln.
MDL Number MFCD00006183
Storage Note 1 2° to 8°C

Specifications

Chemical Name or Material MOPS
CAS 1132-61-2
Name Note low endotoxin
MDL Number MFCD00006183
Accessories

2° to 8°C