missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Montmorillonite K10

$31.42 - $31.42

Chemical Identifiers

CAS 1318-93-0
MDL Number MFCD00132796
Merck Index 14,6260
Chemical Name or Material Montmorillonite K10
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1516030 SDS Alfa Aesar™
L1516030
250g Each for $31.42
Specifications

Description

Applications
Used for support for catalysts, preparation of clay nanopaper composites of nacre-like structure, conversion of triose sugars with alcohols to alkyl lactates and Deprotection of ketone dimethylhydrazones.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

1318-93-0
MFCD00132796
14,6260
Montmorillonite K10

Specifications

1318-93-0
14,6260
Odorless
MFCD00132796
250g
Montmorillonite K10

Safety and Handling

RTECSNumber : CT9450000