MilliporeSigma™ Montage™ SEQ96 Sequencing Reaction Cleanup Kit
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
Supplier:  MilliporeSigma™ LSKS09604
 View more versions of this product
Catalog No. LSKS09604
Description
Description
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
Specifications
Specifications
DNA sequencing
96-well filter plates and Injection solution
4 Pack