missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Monosodium L-Glutamate, FCC, 98.5-101.5%, Spectrum™ Chemical
G1038, 6106-04-3, C5H8NNaO4•H2O
$124.52 - $358.58
Chemical Identifiers
CAS | 6106-04-3 |
---|---|
Molecular Formula | C5H10NNaO5 |
Molecular Weight (g/mol) | 187.13 |
InChI Key | GJBHGUUFMNITCI-QTNFYWBSSA-M |
IUPAC Name | sodium (4S)-4-amino-4-carboxybutanoate hydrate |
SMILES | O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18604683
|
Spectrum Chemical Mfg Cor
G1038100GM |
100 g | Poly Bottle |
Each for $124.52
|
|
||||
18604684
|
Spectrum Chemical Mfg Cor
G1038500GM |
500 g | Poly Bottle |
Each for $165.83
|
|
||||
18604685
|
Spectrum Chemical Mfg Cor
G103825KG |
2.5 kg | Poly Pail |
Each for $358.58
|
|
||||
Description
Spectrum™ Chemical Monosodium L-Glutamate, FCC is used in the food industry as a flavor with an umami taste. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
6106-04-3 | |
187.13 | |
sodium (4S)-4-amino-4-carboxybutanoate hydrate |
C5H10NNaO5 | |
GJBHGUUFMNITCI-QTNFYWBSSA-M | |
O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O |
Specifications
6106-04-3 | |
C5H10NNaO5 | |
0.005 | |
GJBHGUUFMNITCI-QTNFYWBSSA-M | |
sodium (4S)-4-amino-4-carboxybutanoate hydrate | |
98.5 to 101.5% | |
100 g |
1 | |
5 mg/kg | |
Poly Bottle | |
O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O | |
187.13 | |
FCC | |
+24.5° to+25.5° |