missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monooctyl Maleate 95.0+%, TCI America™

Manufacturer:  TCI America M0615500ML

 View more versions of this product

Catalog No. M0615500ML



Specifications

Specifications

Monooctyl Maleate
500mL
C12H20O4
Maleic Acid Mono(2-ethylhexyl) Ester, Mono(2-ethylhexyl) Maleate, Maleic Acid Monooctyl Ester
Clear Liquid at 20°C
Yellow
2370-71-0
MFCD00151510
228.29
≥95.0% (GC)

Chemical Identifiers

2370-71-0
C12H20O4
MFCD00151510
Maleic Acid Mono(2-ethylhexyl) Ester, Mono(2-ethylhexyl) Maleate, Maleic Acid Monooctyl Ester
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1105

TSCA : No

Recommended Storage : Freezer

Documents