missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monooctyl Maleate, TCI America™

$50.42 - $221.53

Chemical Identifiers

CAS 2370-71-0
Molecular Formula C12H20O4
MDL Number MFCD00151510
Synonym Maleic Acid Mono(2-ethylhexyl) Ester, Mono(2-ethylhexyl) Maleate, Maleic Acid Monooctyl Ester
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M0615500ML View Documents TCI America
M0615500ML
500mL Each for $221.53
 
M061525ML View Documents TCI America
M061525ML
25mL Each for $50.42
 
Specifications

Chemical Identifiers

2370-71-0
C12H20O4
MFCD00151510
Maleic Acid Mono(2-ethylhexyl) Ester, Mono(2-ethylhexyl) Maleate, Maleic Acid Monooctyl Ester

Specifications

Yellow
2370-71-0
MFCD00151510
228.29
≥95.0% (GC)
500mL
C12H20O4
Maleic Acid Mono(2-ethylhexyl) Ester, Mono(2-ethylhexyl) Maleate, Maleic Acid Monooctyl Ester
Clear Liquid at 20°C
Monooctyl Maleate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1105

TSCA : No

Recommended Storage : Freezer