missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monomethyl trans-1,4-Cyclohexanedicarboxylate 97.0+%, TCI America™

Manufacturer:  Tci America M252625G

 View more versions of this product

Catalog No. M252625G



Specifications

Specifications

Monomethyl trans-1,4-Cyclohexanedicarboxylate
128°C
15177-67-0
MFCD01311247
ZQJNPHCQABYENK-UHFFFAOYSA-N
4-methoxycarbonylcyclohexane-1-carboxylic acid
296850
Crystal-Powder at 20°C
White
25g
C9H14O4
trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester, Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate
COC(=O)C1CCC(CC1)C(=O)O
186.207
186.21
≥97.0% (GC,T)

Chemical Identifiers

15177-67-0
186.207
ZQJNPHCQABYENK-UHFFFAOYSA-N
296850
COC(=O)C1CCC(CC1)C(=O)O
C9H14O4
MFCD01311247
trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester, Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate
4-methoxycarbonylcyclohexane-1-carboxylic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions