missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monomethyl Suberate, TCI America™

Manufacturer:  TCI America S044025G

 View more versions of this product

Catalog No. S044025G



Specifications

Specifications

Monomethyl Suberate
White-Yellow
3946-32-5
MFCD00004427
KOVPXZDUVJGGFU-UHFFFAOYSA-N
8-methoxy-8-oxooctanoic acid
554191
Crystal-Lump at 20°C
135°C
25g
C9H16O4
suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
COC(=O)CCCCCCC(=O)O
188.223
188.22
≥98.0% (GC,T)

Chemical Identifiers

3946-32-5
188.223
KOVPXZDUVJGGFU-UHFFFAOYSA-N
554191
COC(=O)CCCCCCC(=O)O
C9H16O4
MFCD00004427
suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
8-methoxy-8-oxooctanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents