missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monomethyl 5-Norbornene-2,3-dicarboxylate, TCI America™

$120.70 - $319.31

Chemical Identifiers

CAS 36897-94-6
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00167743
InChI Key JYZKYCYHXBQTCY-UHFFFAOYSA-N
Synonym 5-Norbornene-2,3-dicarboxylic Acid Monomethyl Ester, Methyl Hydrogen 5-Norbornene-2,3-dicarboxylate
PubChem CID 98465
IUPAC Name 3-methoxycarbonylbicyclo[2.2.1]hept-5-ene-2-carboxylic acid
SMILES COC(=O)C1C2CC(C1C(=O)O)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N052525G View Documents TCI America
N052525G
25g Each for $319.31
 
N05255G View Documents TCI America
N05255G
5g Each for $120.70
 
Specifications

Chemical Identifiers

36897-94-6
196.202
JYZKYCYHXBQTCY-UHFFFAOYSA-N
98465
COC(=O)C1C2CC(C1C(=O)O)C=C2
C10H12O4
MFCD00167743
5-Norbornene-2,3-dicarboxylic Acid Monomethyl Ester, Methyl Hydrogen 5-Norbornene-2,3-dicarboxylate
3-methoxycarbonylbicyclo[2.2.1]hept-5-ene-2-carboxylic acid

Specifications

White
25g
C10H12O4
5-Norbornene-2,3-dicarboxylic Acid Monomethyl Ester, Methyl Hydrogen 5-Norbornene-2,3-dicarboxylate
COC(=O)C1C2CC(C1C(=O)O)C=C2
196.202
196.20
≥99.0% (T)
100°C
36897-94-6
MFCD00167743
JYZKYCYHXBQTCY-UHFFFAOYSA-N
3-methoxycarbonylbicyclo[2.2.1]hept-5-ene-2-carboxylic acid
98465
Crystal-Powder at 20°C
Monomethyl 5-Norbornene-2,3-dicarboxylate
Safety and Handling

Safety and Handling

TSCA : Yes