missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monomethyl 2,6-Pyridinedicarboxylate, TCI America™

$174.17 - $594.31

Chemical Identifiers

CAS 7170-36-7
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00806109
InChI Key VWIOMFMPIVMLIR-UHFFFAOYSA-N
Synonym 2,6-Pyridinedicarboxylic Acid Monomethyl Ester, Monomethyl Dipicolinate, Dipicolinic Acid Monomethyl Ester
PubChem CID 4019172
IUPAC Name 6-methoxycarbonylpyridine-2-carboxylic acid
SMILES COC(=O)C1=CC=CC(=N1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25801G View Documents TCI America
M25801G
1g Each for $174.17
 
M25805G View Documents TCI America
M25805G
5g Each for $594.31
 
Specifications

Chemical Identifiers

7170-36-7
181.147
VWIOMFMPIVMLIR-UHFFFAOYSA-N
4019172
COC(=O)C1=CC=CC(=N1)C(=O)O
C8H7NO4
MFCD00806109
2,6-Pyridinedicarboxylic Acid Monomethyl Ester, Monomethyl Dipicolinate, Dipicolinic Acid Monomethyl Ester
6-methoxycarbonylpyridine-2-carboxylic acid

Specifications

White
1g
C8H7NO4
2,6-Pyridinedicarboxylic Acid Monomethyl Ester, Monomethyl Dipicolinate, Dipicolinic Acid Monomethyl Ester
COC(=O)C1=CC=CC(=N1)C(=O)O
181.147
181.15
≥97.0% (GC,T)
153°C
7170-36-7
MFCD00806109
VWIOMFMPIVMLIR-UHFFFAOYSA-N
6-methoxycarbonylpyridine-2-carboxylic acid
4019172
Crystal-Powder at 20°C
Monomethyl 2,6-Pyridinedicarboxylate
Safety and Handling

Safety and Handling

TSCA : No