Please login to your online account to display your discounted pricing

Covidien Monoject™ Corvac™ Silicone-Coated Tubes with Red and Gray Mottled Stoppers

Integrated serum separator tubes

$180.87 - $181.26

Specifications

Color Red and gray
Material Silicone coated; Gel barrier
Sterile Yes
View More Specs
Products
Catalog Number Mfr. No. Tube Size Capacity (Metric) Price Quantity    

22-029-312

 
covidien
8881302015
16 x 100mm 9mL Pack of 100 Each for $180.87
Case of 5 Pack for $904.35

22-029-313

 
covidien
8881302072
13 x 75mm 4mL Pack of 100 Each for $181.26
Case of 5 Pack for $906.30
Description & Specifications

Specifications

Color Red and gray
Material Silicone coated; Gel barrier
Sterile Yes

Improved performance and more sample yield for laboratory procedures requiring serum

  • Contains a positive gel barrier between blood cells and serum
  • Interior silicone glass powder activates the clotting of blood
  • Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging; tubes are sterilized by gamma radiation
  • Crown Corvac™ gel placement for improved performance and more sample yield
  • TDM compatible
  • Allows for sensitive and routine pharmacokinetic testing
  • Plastic coating prevents tube cracking
  • Can be used with both angle-head and horizontal-head centrifuges

  • Laboratory procedures requiring serum with a positive gel barrier between blood cells and serum; general chemistry, serology and toxicology procedures