Please login to your online account to display your discounted pricing

Monoethanolamine, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  931403

 View more versions of this product

Catalog No. 02003497

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 61.08
Chemical Name or Material Monoethanolamine
Molecular Formula HOCH2CH2NH2
CAS 141-43-5
Packaging Narrow Mouth Amber Glass Bottle
Synonym 2-aminoethanol
Specific Gravity 1L = 1.02kg
Flash Point 86.00°C
Quantity 4L

Specifications

Chemical Name or Material Monoethanolamine
CAS 141-43-5
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula HOCH2CH2NH2
Formula Weight 61.08
Synonym 2-aminoethanol