Please login to your online account to display your discounted pricing

Monoethanolamine, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$418.24 - $458.33

Chemical Identifiers

Chemical Name or Material Monoethanolamine
CAS 141-43-5
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula HOCH2CH2NH2
Formula Weight 61.08
Synonym 2-aminoethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003497

 
avantor performance
931403
Narrow Mouth Amber Glass Bottle 4L Each for $458.33

02003498

 
avantor performance
931401
Narrow Mouth Amber Glass Bottle 500mL Each for $418.24
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 61.08
Chemical Name or Material Monoethanolamine
Molecular Formula HOCH2CH2NH2
CAS 141-43-5
Packaging Narrow Mouth Amber Glass Bottle
Synonym 2-aminoethanol
Specific Gravity 1L = 1.02kg
Flash Point 86.00°C
Quantity 4L